Få mere at vide
(4S)-1-(Methyl-d3)-2-oxo-4-imidazolidinecarboxylic Acid, tert-Butyl Ester, TRC
Artikelnummer. 30469990
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30469990 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30469990 Leverandør Toronto Research Chemicals Leverandørnr. TRCM32059710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (4S)-1-(Methyl-d3)-2-oxo-4-imidazolidinecarboxylic Acid','tert-Butyl Ester
CAS 1217704-07-8
Molekylær formel C9H13D3N2O3
Mængde 10 mg
Synonym (4S)-1-(Methyl-d3)-2-oxo-4-imidazolidinecarboxylic Acid 1,1-Dimethylethyl Ester
InChI formel InChI=1S/C9H16N2O3/c1-9(2,3)14-7(12)6-5-11(4)8(13)10-6/h6H,5H2,1-4H3,(H,10,13)/t6-/m0/s1/i4D3
SMIL [2H]C([2H])([2H])N1C[C@H](NC1=O)C(=O)OC(C)(C)C
IUPAC navn tert-butyl (4S)-2-oxo-1-(trideuteriomethyl)imidazolidine-4-carboxylate
Molekylvægt (g/mol) 203.25
Formel vægt 203.1349