Få mere at vide
(4R,6R)-6-(Cyanomethyl)-2,2-dimethyl-1,3-dioxane-4-acetic Acid 1,1-Dimethylethyl Ester, TRC
Artikelnummer. 30354917
Skift visning
Mængde:
500 mg
2.5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30354917 10 g
30366298 500 mg
30361192 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30354917 Leverandør Toronto Research Chemicals Leverandørnr. TRCC98202010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 125971-94-0
Molekylær formel C14 H23 N O4
Molekylvægt (g/mol) 269.34
IUPAC navn tert-butyl 2-[(4R,6R)-6-(cyanomethyl)-2,2-dimethyl-1,3-dioxan-4-yl]acetate
SMIL CC(C)(C)OC(=O)C[C@H]1C[C@@H](CC#N)OC(C)(C)O1
Tekniske data
Kemisk navn eller materiale 1,1-Dimethylethyl 2-[(4R,6R)-6-(Cyanomethyl)-2,2-dimethyl-1,3-dioxan-4-yl]acetate
Mængde 10 g
Synonym 1,3-Dioxane-4-acetic acid, 6-(cyanomethyl)-2,2-dimethyl-, 1,1-dimethylethyl ester, (4R,6R)-, (4R,6R)-6-(Cyanomethyl)-2,2-dimethyl-1,3-dioxane-4-acetic Acid 1,1-Dimethylethyl Ester, : 1,3-Dioxane-4-acetic acid, 6-(cyanomethyl)-2,2-dimethyl-, 1,1-dimethylethyl ester, (4R-cis)-, ((4R,6R)-6-(Cyanomethyl)-2,2-dimethyl-[1,3]dioxan-4-yl)acetic acid tert-butyl ester, (6R-Cyanomethyl-2,2-dimethyl-[1,3]dioxan-4R-yl)acetic acid tert-butyl ester
InChI formel InChI=1S/C14H23NO4/c1-13(2,3)19-12(16)9-11-8-10(6-7-15)17-14(4,5)18-11/h10-11H,6,8-9H2,1-5H3/t10-,11-/m1/s1
Formel vægt 269.1627