Få mere at vide
(4R)-4-(Isopropylamino)-2-(3-methoxypropyl)-3,4-dihydro-2H-thieno[3,2-e]-1,2-thiazine-6-sulfonamide 1,1-Dioxide, Mikromol™
Artikelnummer. 30050661
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050661 25 mg
17901576 100 mg
30464235 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30050661 Leverandør Mikromol Leverandørnr. MM1300.050025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(4R)-4-(Isopropylamino)-2-(3-methoxypropyl)-3,4-dihydro-2H-thieno[3,2-e]-1,2-thiazine-6-sulfonamide 1,1-Dioxide
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (4R)-4-(Isopropylamino)-2-(3-methoxypropyl)-3,4-dihydro-2H-thieno[3,2-e]-1,2-thiazine-6-sulfonamide-1,1-dioxide
CAS 2985481-46-5
Analyt- eller komponentnavne (4R)-4-(Isopropylamino)-2-(3-methoxypropyl)-3,4-dihydro-2H-thieno[3,2-e]-1,2-thiazine-6-sulfonamide-1,1-dioxide
Forsendelsestilstand Room Temperature
Molekylær formel C13 H23 N3 O5 S3
Certificeringer/compliance ISO 17025
Synonym (4R)-4-(Isopropylamino)-2-(3-methoxypropyl)-1,1-dioxo-3,4-dihydrothieno[3,2-e]thiazine-6-sulfonamide, (4R)-4-(Isopropylamino)-2-(3-methoxypropyl)-3,4-dihydro-2H-thieno[3,2-e]-1,2-thiazine-6-sulfonamide-1,1-dioxide, N-Desethyl N-Isopropyl Brinzolamide
InChI formel InChI=1S/C13H23N3O5S3/c1-9(2)15-11-8-16(5-4-6-21-3)24(19,20)13-10(11)7-12(22-13)23(14,17)18/h7,9,11,15H,4-6,8H2,1-3H3,(H2,14,17,18)/t11-/m0/s1
SMIL COCCCN1C[C@H](NC(C)C)c2cc(sc2S1(=O)=O)S(=O)(=O)N
IUPAC navn (4R)-2-(3-methoxypropyl)-1,1-dioxo-4-(propan-2-ylamino)-3,4-dihydrothieno[3,2-e]thiazine-6-sulfonamide
Molekylvægt (g/mol) 397.53
Mængde 25 mg
Formel vægt 397.08 g/mol