Få mere at vide
(4R)-4-(1-Methylethyl)-2-cyclohexen-1-one (~80% ee), TRC
Artikelnummer. 30473469
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30473469 10 mg
30469078 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30473469 Leverandør Toronto Research Chemicals Leverandørnr. TRCM22386510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (4R)-4-(1-Methylethyl)-2-cyclohexen-1-one
CAS 2158-59-0
Molekylær formel C9H14O
Mængde 10 mg
Synonym (R)-4- Isopropylcyclohex-2-enone, (R)-4-(1-methylethyl)-2-cyclohexen-1-one, (R)-(-)- (8CI)-4-isopropyl-2-cyclohexen-1-one, (4R)-4-(1-Methylethyl)-2-cyclohexen-1-one, (R)-(-)-Cryptone, l-Cryptone
InChI formel InChI=1S/C9H14O/c1-7(2)8-3-5-9(10)6-4-8/h3,5,7-8H,4,6H2,1-2H3/t8-/m1/s1
SMIL CC(C)[C@H]1CCC(=O)C=C1
IUPAC navn (4R)-4-propan-2-ylcyclohex-2-en-1-one
Molekylvægt (g/mol) 138.21
Formel vægt 138.1045