missing translation for 'onlineSavingsMsg'
Få mere at vide

(4R)-1-Methyl-4-propyl-L-proline Methyl Ester, TRC

Artikelnummer. 30464903
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30464903 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30464903 Leverandør Toronto Research Chemicals Leverandørnr. TRCM32029025MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale (4R)-1-Methyl-4-propyl-L-proline Methyl Ester
CAS 13380-39-7
Molekylær formel C10H19NO2
Mængde 25 mg
Synonym 4-Propyl-hygric Acid Methyl Ester, (4R)-4-Propyl-hygric Acid Methyl Ester, (2S,4R)-4-Propyl-1-methylpyrrolidine-2-carboxylic Acid Methyl Ester, trans-4-Propyl-1methyl-L-proline Methyl Ester
InChI formel InChI=1S/C10H19NO2/c1-4-5-8-6-9(10(12)13-3)11(2)7-8/h8-9H,4-7H2,1-3H3/t8-,9+/m1/s1
SMIL CCC[C@@H]1C[C@H](N(C)C1)C(=O)OC
IUPAC navn methyl (2S,4R)-1-methyl-4-propylpyrrolidine-2-carboxylate
Molekylvægt (g/mol) 185.26
Formel vægt 185.1416
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.