missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4EBP1 Antibody (007), DyLight 680, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Bio-Techne NBP2-89207FR
This item is not returnable.
View return policy
Description
4EBP1 Monoclonal specifically detects 4EBP1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
4EBP1 | |
Monoclonal | |
DyLight 680 | |
4E-BP1, eIF4E-binding protein 1, eukaryotic translation initiation factor 4E binding protein 1,4EBP1,4E-BP1BP-1, eukaryotic translation initiation factor 4E-binding protein 1, PHAS-IMGC4316, Phosphorylated heat- and acid-stable protein regulated by insulin 1 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human 4EBP1 (NP_004086.1; Ser2-Ile118). | |
0.1 mL | |
Cancer, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Lipid and Metabolism, mTOR Pathway, Phospho Specific, Stem Cell Markers | |
1978 | |
Store at 4°C in the dark. | |
IgG |
Immunofluorescence | |
7 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction