missing translation for 'onlineSavingsMsg'
Learn More

(4E)-5-(4-Methoxy-d3-2,3,6-trimethylphenyl)-3-methyl-2,4-pentadienal, TRC

Product Code. 30469880
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Mængde:
2.5 mg
missing translation for 'unitSize'
2.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde unitSize
30469880 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30469880 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCM2716022.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale (4E)-5-(4-Methoxy-d3-2,3,6-trimethylphenyl)-3-methyl-2,4-pentadienal
CAS 1185247-91-9
Molekylær formel C16H17D3O2
Mængde 2.5 mg
InChI formel InChI=1S/C16H20O2/c1-11(8-9-17)6-7-15-12(2)10-16(18-5)14(4)13(15)3/h6-10H,1-5H3/b7-6+,11-8+/i5D3
SMIL [2H]C([2H])([2H])Oc1cc(C)c(\C=C\C(=C\C=O)\C)c(C)c1C
IUPAC navn (2E,4E)-3-methyl-5-[2,3,6-trimethyl-4-(trideuteriomethoxy)phenyl]penta-2,4-dienal
Molekylvægt (g/mol) 247.35
Formel vægt 247.1652
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.