Få mere at vide
4beta-Hydroxy Cholesterol-d7 4-Acetate, TRC
Artikelnummer. 30406172
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30406172 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30406172 Leverandør Toronto Research Chemicals Leverandørnr. TRCH9179871MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4beta-Hydroxy Cholesterol-d7 4-Acetate
CAS 1363529-54-7
Molekylær formel C29 D7 H41 O3
Mængde 1 mg
Synonym (3β,4β)-Cholest-5-ene-3,4-diol-d7 4-Acetate, 4β-Acetoxycholest-5-en-3β-ol-d7
InChI formel InChI=1S/C29H48O3/c1-18(2)8-7-9-19(3)22-12-13-23-21-10-11-25-27(32-20(4)30)26(31)15-17-29(25,6)24(21)14-16-28(22,23)5/h11,18-19,21-24,26-27,31H,7-10,12-17H2,1-6H3/t19-,21+,22-,23+,24+,26+,27-,28-,29-/m1/s1/i1D3,2D3,18D
SMIL [2H]C([2H])([2H])C([2H])(CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4[C@@H](OC(=O)C)[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C)C([2H])([2H])[2H]
IUPAC navn [(3S,4R,8S,9S,10R,13R,14S,17R)-3-hydroxy-10,13-dimethyl-17-[(2R)-6,7,7,7-tetradeuterio-6-(trideuteriomethyl)heptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-4-yl] acetate
Molekylvægt (g/mol) 451.733
Formel vægt 451.404