missing translation for 'onlineSavingsMsg'
Learn More

4-Vinylbenzyl chloride, 90%, technical

Product Code. p-3737737 Shop All Thermo Scientific Chemicals Products
25 g, Glass bottle, 25g
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 1592-20-7 | C9H9Cl | 152.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1592-20-7
Molecular Formula C9H9Cl
Molecular Weight (g/mol) 152.62
MDL Number MFCD00051362
InChI Key ZRZHXNCATOYMJH-UHFFFAOYSA-N
Synonym 4-vinylbenzyl chloride, 1-chloromethyl-4-vinylbenzene, 4-chloromethylstyrene, 4-vinylbenzylchloride, benzene, 1-chloromethyl-4-ethenyl, 1-chloromethyl-4-ethenylbenzene, 4-chloromethyl styrene, p-vinylbenzyl chloride, unii-cjl4oj6bs0, cjl4oj6bs0
PubChem CID 74126
IUPAC Name 1-(chloromethyl)-4-ethenylbenzene
SMILES ClCC1=CC=C(C=C)C=C1

Specifications

CAS Min % 88.0
CAS Max % 100.0
Color Yellow
Density 1.0800g/mL
Flash Point 105°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula CH2CHC6H4CH2Cl
Refractive Index 1.5731 to 1.5751
Quantity 25 g
Specific Gravity 1.08
Stabilizer about 500ppm 4-tert-Butylcatechol, about 500ppm Nitrocresol + Nitrophenol
Formula Weight 152.62
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material 4-Vinylbenzyl chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Vinylbenzyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
  • P331-Do NOT induce vomiting.
  • P363-Wash contaminated clothing before reuse.
  • P403-Store in a well-ventilated place.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : 4-Vinylbenzyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Vinylbenzyl chloride, 90%, technical >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.