missing translation for 'onlineSavingsMsg'
Learn More

4-[(Trimethylsilyl)ethynyl]benzaldehyde, 97%, Thermo Scientific Chemicals

Catalog Number 15441188
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
77123-57-0
C12H14OSi
202.33
MFCD02093765
UZQDUXAJFTWMDT-UHFFFAOYSA-N
4-trimethylsilyl ethynyl benzaldehyde, 4-trimethylsilyl ethynylbenzaldehyde, 4-2-trimethylsilylethynyl benzaldehyde, 4-2-trimethylsilyl ethynyl benzaldehyde, benzaldehyde, 4-trimethylsilyl ethynyl, acmc-1bios, p-trimethylsilylethynylbenzaldehyde, 4-trimethylsilylethynylbenzaldehyde
2771643
4-(2-trimethylsilylethynyl)benzaldehyde
C[Si](C)(C)C#CC1=CC=C(C=O)C=C1
This item is not returnable. View return policy
77123-57-0
C12H14OSi
202.33
MFCD02093765
UZQDUXAJFTWMDT-UHFFFAOYSA-N
4-trimethylsilyl ethynyl benzaldehyde, 4-trimethylsilyl ethynylbenzaldehyde, 4-2-trimethylsilylethynyl benzaldehyde, 4-2-trimethylsilyl ethynyl benzaldehyde, benzaldehyde, 4-trimethylsilyl ethynyl, acmc-1bios, p-trimethylsilylethynylbenzaldehyde, 4-trimethylsilylethynylbenzaldehyde
2771643
4-(2-trimethylsilylethynyl)benzaldehyde
C[Si](C)(C)C#CC1=CC=C(C=O)C=C1

It is an important raw material and intermediate used in organic synthesis, Pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, Pharmaceuticals, agrochemicals and dyestuffs.

Notes
Store at 4°C. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 66°C to 70°C
Quantity 5 g
Formula Weight 202.33
Percent Purity 97%
Chemical Name or Material 4-[(Trimethylsilyl)ethynyl]benzaldehyde

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'