missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)thiophenol, 97%, Thermo Scientific Chemicals

Catalog Number p-7020684
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
825-83-2
C7H5F3S
178.172
MFCD00042320
WCMLRSZJUIKVCW-UHFFFAOYSA-N
4-trifluoromethyl thiophenol, 4-trifluoromethyl benzenethiol, 4-trifluoromethyl benzene-1-thiol, 4-trifluoromethylthiophenol, 4-mercaptobenzotrifluoride, benzenethiol, 4-trifluoromethyl, 4-trifluoromethyl t, acmc-20amtl, pubchem6864
136653
4-(trifluoromethyl)benzenethiol
C1=CC(=CC=C1C(F)(F)F)S
This item is not returnable. View return policy
825-83-2
C7H5F3S
178.172
MFCD00042320
WCMLRSZJUIKVCW-UHFFFAOYSA-N
4-trifluoromethyl thiophenol, 4-trifluoromethyl benzenethiol, 4-trifluoromethyl benzene-1-thiol, 4-trifluoromethylthiophenol, 4-mercaptobenzotrifluoride, benzenethiol, 4-trifluoromethyl, 4-trifluoromethyl t, acmc-20amtl, pubchem6864
136653
4-(trifluoromethyl)benzenethiol
C1=CC(=CC=C1C(F)(F)F)S

4-(Trifluoromethyl)thiophenol is used an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Trifluoromethyl)thiophenol is used an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store in cool dry place in tightly closed container. With good ventilation. Store away from air and oxidizing agent.
TRUSTED_SUSTAINABILITY
Density 1.3
Boiling Point 66°C to 68°C (15 mmHg)
Odor Stench
Refractive Index 1.492
Quantity 1 g
UN Number UN2810
Beilstein 2045665
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 178.18
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethyl)thiophenol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'