Learn More
4-(Trifluoromethyl)phenylacetonitrile, 98%, BLD Pharm™
Product Code. 30948661
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
500 g
Unit Size:
100g
10g
25g
500g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30948661 100 g 100g
30947076 10 g 10g
30944499 500 g 500g
30943001 25 g 25g
30931667 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 30948661 Supplier BLD Pharm Supplier No. BD11504100g
 Restricted Item
Forventet forsendelse: 27-11-2026
Specifications
CAS 2338-75-2
Molecular Formula C9H6F3N
MDL Number MFCD00001921
Quantity 100 g
SMILES FC(F)(F)C1=CC=C(CC#N)C=C1
Molecular Weight (g/mol) 185.15
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethyl)phenylacetonitrile