missing translation for 'onlineSavingsMsg'
Learn More
4-(Trifluoromethyl)phenol, 98% Product Code.: 10763561

4-(Trifluoromethyl)phenol, 98%

Product Code. p-3779731 Shop All Thermo Scientific Chemicals Products
1 g, Glass bottle, 1g
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

Product Code. 10763561

Brand: Thermo Scientific Acros 176440010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

CAS: 402-45-9 | C7H5F3O | 162.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 402-45-9
Molecular Formula C7H5F3O
Molecular Weight (g/mol) 162.11
MDL Number MFCD00002363
InChI Key BAYGVMXZJBFEMB-UHFFFAOYSA-N
Synonym 4-trifluoromethyl phenol, 4-hydroxybenzotrifluoride, alpha,alpha,alpha-trifluoro-p-cresol, p-trifluoromethylphenol, p-hydroxybenzotrifluoride, phenol, 4-trifluoromethyl, a,a,a-trifluoro-p-cresol, 4-trifluoromethyl-phenol, para-trifluoromethylphenol
PubChem CID 67874
ChEBI CHEBI:42578
IUPAC Name 4-(trifluoromethyl)phenol
SMILES C1=CC(=CC=C1C(F)(F)F)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 40.0°C to 46.0°C
Density 83g/mL
Boiling Point 71.5°C to 72.0°C (8.0 mmHg)
Flash Point 83°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CF3C6H4OH
Quantity 1 g
Formula Weight 162.11
Percent Purity 98%
Physical Form Crystals
Chemical Name or Material 4-(Trifluoromethyl)phenol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-(Trifluoromethyl)phenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H301-Toxic if swallowed.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-(Trifluoromethyl)phenol, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.