Få mere at vide
4-(Trifluoromethyl)nicotinoyl Glycine, TRC
Artikelnummer. 17974085
Skift visning
Mængde:
50 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17974085 1 g
17913945 250 mg
17963905 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17974085 Leverandør Toronto Research Chemicals Leverandørnr. TRCT7913751G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 207502-65-6
Molekylær formel C9 H7 F3 N2 O3
Molekylvægt (g/mol) 248.16
Synonym Glycine, N-[[4-(trifluoromethyl)-3-pyridinyl]carbonyl]-, 4-(Trifluoromethyl)nicotinoyl glycine, N-[[4-(Trifluoromethyl)-3-pyridinyl]carbonyl]glycine, 2-(4-(Trifluoromethyl)nicotinamido)acetic acid, 4-(Trifluoromethyl)nicotinoyl glycine, TFNG, Flonicamid-carboxylic Acid, Flonicamid-carboxylic acid
IUPAC navn 2-[[4-(trifluoromethyl)pyridine-3-carbonyl]amino]acetic acid
SMIL OC(=O)CNC(=O)c1cnccc1C(F)(F)F
Tekniske data
Kemisk navn eller materiale 4-(Trifluoromethyl)nicotinoyl Glycine
Renhedsgrad noter HPLC
InChI formel InChI=1S/C9H7F3N2O3/c10-9(11,12)6-1-2-13-3-5(6)8(17)14-4-7(15)16/h1-3H,4H2,(H,14,17)(H,15,16)
Mængde 1 g
Formel vægt 248.0409
Procent renhed >95%