Få mere at vide
4-(Trifluoromethyl)nicotinamide, TRC
Artikelnummer. 30134444
Skift visning
Mængde:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134444 25 g
30132972 5 g
17944025 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30134444 Leverandør Toronto Research Chemicals Leverandørnr. TRCT79131525G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 158062-71-6
Molekylær formel C7 H5 F3 N2 O
Molekylvægt (g/mol) 190.12
Synonym 4-(Trifluoromethyl)-3-pyridinecarboxamide, 4-Trifluoromethylnicotinamide, TFNA-AM
IUPAC navn 4-(trifluoromethyl)pyridine-3-carboxamide
SMIL NC(=O)c1cnccc1C(F)(F)F
Tekniske data
Kemisk navn eller materiale 4-(Trifluoromethyl)nicotinamide
Mængde 25 g
InChI formel InChI=1S/C7H5F3N2O/c8-7(9,10)5-1-2-12-3-4(5)6(11)13/h1-3H,(H2,11,13)
Formel vægt 190.0354