Learn More
4-(Trifluoromethyl)benzenesulfonyl chloride, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 2991-42-6 |
| Molecular Formula | C7H4ClF3O2S |
| MDL Number | MFCD00042422 |
| Quantity | 10 g |
| SMILES | FC(C1=CC=C(C=C1)[S](=O)(=O)Cl)(F)F |
| Molecular Weight (g/mol) | 244.62 |
| Percent Purity | 97% |
| Chemical Name or Material | 4-(Trifluoromethyl)benzenesulfonyl chloride |