missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Trifluoromethyl)benzene-1-carboximidamide hydrochloride dihydrate, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
10 g
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Linear Formula | HCl·H2O |
Quantity | 1 g |
IUPAC Name | 4-(trifluoromethyl)benzenecarboximidamide;dihydrate;hydrochloride |
Formula Weight | 242.63 |
Percent Purity | 97% |
Chemical Name or Material | 4-(Trifluoromethyl)benzene-1-carboximidamide hydrochloride hydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'