missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)anisole, 98%, Thermo Scientific Chemicals

Catalog Number p-7055575
142.58 DKK valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
402-52-8
C8H7F3O
176.138
MFCD03094456
CFIPQRIPCRRISV-UHFFFAOYSA-N
4-trifluoromethyl anisole, 1-methoxy-4-trifluoromethyl benzene, 4-trifluoromethylanisole, benzene, 1-methoxy-4-trifluoromethyl, 4-methoxybenzotrifluoride, buttpark 180\07-83, p-trifluoromethylanisole, pubchem10126, 4-trifluoromethyl-anisol, 1-methoxy-4-trifluoromethyl-benzene
600604
1-methoxy-4-(trifluoromethyl)benzene
COC1=CC=C(C=C1)C(F)(F)F
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
402-52-8
C8H7F3O
176.138
MFCD03094456
CFIPQRIPCRRISV-UHFFFAOYSA-N
4-trifluoromethyl anisole, 1-methoxy-4-trifluoromethyl benzene, 4-trifluoromethylanisole, benzene, 1-methoxy-4-trifluoromethyl, 4-methoxybenzotrifluoride, buttpark 180\07-83, p-trifluoromethylanisole, pubchem10126, 4-trifluoromethyl-anisol, 1-methoxy-4-trifluoromethyl-benzene
600604
1-methoxy-4-(trifluoromethyl)benzene
COC1=CC=C(C=C1)C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -9°C
Color Colorless to Yellow
Density 1.245 g/mL
Boiling Point 169°C (754 mmHg)
Flash Point 60°C (140°F)
Refractive Index 1.445
Quantity 250 mg
UN Number UN1993
Formula Weight 176.14
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-(Trifluoromethyl)anisole
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'