Få mere at vide
4-(Trifluoromethyl)aniline-2,3,5,6-d4, CDN
Artikelnummer. 30320490
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30320490 0.25 g -
30313796 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320490 Leverandør CDN Leverandørnr. CDND56060.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-(Trifluoromethyl)aniline ring-d4
CAS 1219795-48-8
Molekylær formel C7 2H4 H2 F3 N
Mængde 0.25 g
InChI formel InChI=1 S/C7H6F3N/c8-7(9,10)5-1-3-6(11)4-2-5/h1-4 H,11H2/i1D,2 D,3 D,4 D
SMIL [2 H]c1c([2 H])c(c([2 H])c([2 H])c1N)C(F)(F)F
IUPAC navn 2,3,5,6-tetradeuterio-4-(trifluoromethyl)aniline
Molekylvægt (g/mol) 165.15
Formel vægt 165.0703 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity