Få mere at vide
4'-(Trifluoromethyl)-3-biphenylcarboxylic Acid, TRC
Artikelnummer. 30006689
Skift visning
Mængde:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30006689 1 g
30005914 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30006689 Leverandør Toronto Research Chemicals Leverandørnr. TRCT7790951G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 199528-28-4
Molekylær formel C14 H9 F3 O2
Molekylvægt (g/mol) 266.215
Synonym 4'-(Trifluoromethyl)[1,1'-biphenyl]-3-carboxylic Acid, 3-(4-Trifluoromethylphenyl)benzoic Acid;
IUPAC navn 3-[4-(trifluoromethyl)phenyl]benzoic acid
SMIL OC(=O)c1cccc(c1)c2ccc(cc2)C(F)(F)F
Tekniske data
Kemisk navn eller materiale 4'-(Trifluoromethyl)-3-biphenylcarboxylic Acid
Mængde 1 g
InChI formel InChI=1S/C14H9F3O2/c15-14(16,17)12-6-4-9(5-7-12)10-2-1-3-11(8-10)13(18)19/h1-8H,(H,18,19)
Formel vægt 266.055