missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-(Trifluoromethoxy)phenylacetonitrile, 98%

Brand:  Alfa Aesar™ B20267.06

Additional Details : CAS Number : 49561-96-8 Weight : 0.00500kg Transport : UN number : 3439 Chem class : 6.1 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 11499756

  • 1286.00 DKK / 5g

Estimated Shipment
28-05-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

49561-96-8
201.148
LYFCAROXYJTUQF-UHFFFAOYSA-N
142676
C1=CC(=CC=C1CC#N)OC(F)(F)F
C9H6F3NO
MFCD00061240
4-trifluoromethoxy phenylacetonitrile, 2-4-trifluoromethoxy phenyl acetonitrile, 4-trifluoromethoxybenzyl cyanide, 4-trifluoromethoxy benzyl cyanide, benzeneacetonitrile, 4-trifluoromethoxy, 4-trifluoromethoxy-phenyl-acetonitrile, 4-trifluoromethoxy phenyl acetonitrile, 4-trifluoromethoxybenzylcyanide, 1-4-trifluoromethoxy phenyl acetonitrile
2-[4-(trifluoromethoxy)phenyl]acetonitrile

Specifications

4-(Trifluoromethoxy)phenylacetonitrile
>110°C (230°F)
28°C to 30°C
49561-96-8
MFCD00061240
2643789
LYFCAROXYJTUQF-UHFFFAOYSA-N
2-[4-(trifluoromethoxy)phenyl]acetonitrile
142676
98%
1.28
1.448
5g
C9H6F3NO
UN3439
4-trifluoromethoxy phenylacetonitrile, 2-4-trifluoromethoxy phenyl acetonitrile, 4-trifluoromethoxybenzyl cyanide, 4-trifluoromethoxy benzyl cyanide, benzeneacetonitrile, 4-trifluoromethoxy, 4-trifluoromethoxy-phenyl-acetonitrile, 4-trifluoromethoxy phenyl acetonitrile, 4-trifluoromethoxybenzylcyanide, 1-4-trifluoromethoxy phenyl acetonitrile
C1=CC(=CC=C1CC#N)OC(F)(F)F
201.148
201.15
Videos
SDS Documents