missing translation for 'onlineSavingsMsg'
Learn More
4-(Trifluoromethoxy)aniline, 98% Product Code.: 11453427

4-(Trifluoromethoxy)aniline, 98%

Product Code. p-7023543
5 g, 5g
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

Product Code. 11453427

Brand: Thermo Scientific Alfa Aesar A17208.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

CAS: 461-82-5 | C7H6F3NO | 177.126 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 461-82-5
Molecular Formula C7H6F3NO
Molecular Weight (g/mol) 177.126
MDL Number MFCD00041314
InChI Key XUJFOSLZQITUOI-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy aniline, 4-trifluoromethoxy-phenylamine, 4-trifluoromethoxy benzenamine, p-trifluoromethoxyaniline, 4-trifluoroethoxy aniline, benzenamine, 4-trifluoromethoxy, unii-p40l42cvf6, alpha,alpha,alpha-trifluoro-p-anisidine, p-aminotrifluoromethoxybenzene, 4-amino-alpha,alpha,alpha-trifluoroanisole
PubChem CID 600848
IUPAC Name 4-(trifluoromethoxy)aniline
SMILES C1=CC(=CC=C1N)OC(F)(F)F

Specifications

Density 1.31
Boiling Point 73°C to 75°C (10 mmHg)
Flash Point 81°C (178°F)
Odor Characteristic
Refractive Index 1.463
Quantity 5 g
UN Number UN2810
Beilstein 2090209
Formula Weight 177.13
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethoxy)aniline
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot Number10220290 Certificate TypeCertificate of Analysis Date30/08/2024 Product CodeA17208.06,A17208.14
Lot Number10220290 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeA17208.06,A17208.14
Lot Number10196558 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeA17208.06,A17208.14
Lot Number10177421 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeA17208.06,A17208.14
Lot Number10098738 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeA17208.06,A17208.14
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H373-May cause damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P362+P364-Take off contaminated clothing and wash it before reuse.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-(Trifluoromethoxy)aniline, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.