Få mere at vide
4-Thiophenyl Phenyl Diphenyl Sulfonium Hexafluoroantimonate, TRC
Artikelnummer. 30452062
Skift visning
Mængde:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30452062 25 g
30456847 1 g
30451296 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30452062 Leverandør Toronto Research Chemicals Leverandørnr. TRCT36817025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 71449-78-0
Molekylær formel C24 H19 S2 • F6 Sb
Molekylvægt (g/mol) 607.28808
Synonym (OC-6-11)-Diphenyl[4-(phenylthio)phenyl]-sulfonium Hexafluoroantimonate, (OC-6-11)-Hexafluoro-antimonate Diphenyl[4-(phenylthio)phenyl]sulfonium, (4-Thiophenoxyphenyl)diphenylsulfonium hexafluoroantimonate, (p-Thiophenoxyphenyl)diphenylsulfonium hexafluoroantimonate, 4-(Phenylthio)phenyldiphenylsulfonium hexafluoroantimonate, Aceto PI 6976, CPI 100A, CPI 101A, CPI 110A, DTS 103, Diphenyl(4-phenylthiophenyl)sulfonium Hexafluoroantimonate, Diphenyl(p-thiophenoxyphenyl)sulfonium Hexafluoroantimonate, Diphenyl[p-(phenylthio)phenyl]sulfonium Hexafluoroantimonate, PAG 201, S,S-Diphenyl-S-4-thiophenoxyphenylsulfoniumhexafluoroantimonate
IUPAC navn (thiobis(4,1-phenylene))bis(diphenylsulfonium) diphenyl(4-(phenylthio)phenyl)sulfonium hexafluorostibate(V)
SMIL [F-][Sb+5]([F-])([F-])([F-])([F-])[F-].S(C=1C=CC=CC1)C2=CC=C(C=C2)[S+](C=3C=CC=CC3)C=4C=CC=CC4
Tekniske data
Kemisk navn eller materiale 4-Thiophenyl Phenyl Diphenyl Sulfonium Hexafluoroantimonate
Mængde 25 g
InChI formel InChI=1S/C24H19S2.6FH.Sb/c1-4-10-20(11-5-1)25-21-16-18-24(19-17-21)26(22-12-6-2-7-13-22)23-14-8-3-9-15-23;;;;;;;/h1-19H;6*1H;/q+1;;;;;;;+5/p-6
Formel vægt 605.9871