Få mere at vide
4-tert-Octylphenol-glucuronide, TRC
Artikelnummer. 30137351
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137351 1 g
30012473 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30137351 Leverandør Toronto Research Chemicals Leverandørnr. TRCO2938451G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 309257-29-2
Molekylær formel C20H30O7
Molekylvægt (g/mol) 382.45
Synonym beta-D-Glucopyranosiduronic acid 4-(1,1,3,3-Tetramethylbutyl)phenyl
IUPAC navn (2S,3S,6S)-3,4,5-trihydroxy-6-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]oxane-2-carboxylic acid
SMIL CC(C)(C)CC(C)(C)c1ccc(O[C@@H]2O[C@@H]([C@@H](O)C(O)C2O)C(=O)O)cc1
Tekniske data
Kemisk navn eller materiale 4-tert-Octylphenol-glucuronide
Mængde 1 g
InChI formel InChI=1S/C20H30O7/c1-19(2,3)10-20(4,5)11-6-8-12(9-7-11)26-18-15(23)13(21)14(22)16(27-18)17(24)25/h6-9,13-16,18,21-23H,10H2,1-5H3,(H,24,25)/t13?,14-,15?,16-,18+/m0/s1
Formel vægt 382.1992