Få mere at vide
4-tert-Butylphenol-d9 (Major), TRC
Artikelnummer. 30338699
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30338699 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30338699 Leverandør Toronto Research Chemicals Leverandørnr. TRCB69410450MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-tert-Butylphenol-d9
CAS 358730-87-7
Molekylær formel C10H5D9O
Mængde 50 mg
Synonym 4-(1,1-Dimethylethyl)-phenol-d9, p-tert-butyl-phenoll-d9, 4-(1,1-Dimethylethyl)phenol-d9
InChI formel InChI=1S/C10H14O/c1-10(2,3)8-4-6-9(11)7-5-8/h4-7,11H,1-3H3/i1D3,2D3,3D3
SMIL OC1=CC=C(C(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])C=C1
IUPAC navn 4-(2-(methyl-d3)propan-2-yl-1,1,1,3,3,3-d6)phenol
Molekylvægt (g/mol) 159.27
Formel vægt 159.19