Få mere at vide
4-tert-Butyl-d9-phenol-2,3,5,6-d4, TRC
Artikelnummer. 30340803
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30340803 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30340803 Leverandør Toronto Research Chemicals Leverandørnr. TRCB6941065MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-tert-Butyl-d9-phenol-2,3,5,6-d4
CAS 225386-58-3
Molekylær formel C10 D13 H O
Mængde 5 mg
Synonym Phen-2,3,5,6-d4-ol, 4-[1,1-di(methyl-d3)ethyl-2,2,2-d3]- (9CI)
InChI formel InChI=1S/C10H14O/c1-10(2,3)8-4-6-9(11)7-5-8/h4-7,11H,1-3H3/i1D3,2D3,3D3,4D,5D,6D,7D
SMIL [2H]c1c([2H])c(c([2H])c([2H])c1O)C(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn 2,3,5,6-tetradeuterio-4-[1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-yl]phenol
Molekylvægt (g/mol) 163.3
Formel vægt 163.1861