Få mere at vide
4-tert-Butyl-2,6-dimethyl-3-hydroxyphenylacetic Acid, TRC
Artikelnummer. 30136190
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136190 1 g
17991485 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30136190 Leverandør Toronto Research Chemicals Leverandørnr. TRCB6920101G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 55699-12-2
Molekylær formel C14 H20 O3
Molekylvægt (g/mol) 236.31
Synonym [4-(1,1-Dimethylethyl)-3-hydroxy-2,6-dimethylphenyl]acetic Acid, Oxymetazoline Hydrochloride Imp. D (EP)
IUPAC navn 2-(4-tert-butyl-3-hydroxy-2,6-dimethylphenyl)acetic acid
SMIL Cc1cc(c(O)c(C)c1CC(=O)O)C(C)(C)C
Tekniske data
Kemisk navn eller materiale (4-(1,1-Dimethylethyl)-3-hydroxy-2,6-dimethylphenyl)acetic Acid
Mængde 1 g
InChI formel InChI=1S/C14H20O3/c1-8-6-11(14(3,4)5)13(17)9(2)10(8)7-12(15)16/h6,17H,7H2,1-5H3,(H,15,16)
Formel vægt 236.1412