Learn More
Alfa Aesar™ 4-(tert-Butoxycarboxy)benzeneboronic acid, 97%
Click to view available options
Quantity:
250 mg
1 g
Specifications
Melting Point | 140°C to 146°C |
Quantity | 250 mg |
IUPAC Name | [4-[(2-methylpropan-2-yl)oxycarbonyloxy]phenyl]boronic acid |
Formula Weight | 238.05 |
Percent Purity | 97% |
Chemical Name or Material | 4-(tert-Butoxycarboxy)benzeneboronic acid |