Få mere at vide
4-[(RS)-6-Carboxy-9-fluoro-3-methyl-7-oxo-2,3-dihydro-7H-pyrido[1,2,3-de]-1,4-benzoxazine-10-yl]-1-methylpiperazine 1-Oxide Hydrochloride (Ofloxacin N-Oxide Hydrochloride), Mikromol™
Beskrivelse
4-[(RS)-6-Carboxy-9-fluoro-3-methyl-7-oxo-2,3-dihydro-7H-pyrido[1,2,3-de]-1,4-benzoxazine-10-yl]-1-methylpiperazine 1-Oxide Hydrochloride (Ofloxacin N-Oxide Hydrochloride)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Ofloxacin N-Oxide Hydrochloride |
| CAS | 2731368-77-5 |
| Analyt- eller komponentnavne | Ofloxacin N-Oxide Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C18 H20 F N3 O5 . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Ofloxacin Imp. F (EP), 4-[(RS)-6-Carboxy-9-fluoro-3-methyl-7-oxo-2,3-dihydro-7H-pyrido[1,2,3-de]-1,4-benzoxazine-10-yl]-1-methylpiperazine 1-Oxide Hydrochloride, Ofloxacin Imp. F (EP) as Hydrochloride, Ofloxacin N-Oxide Hydrochloride, Ofloxacin Impurity F as Hydrochloride |
| InChI formel | InChI=1S/C18H20FN3O5.ClH/c1-10-9-27-17-14-11(16(23)12(18(24)25)8-21(10)14)7-13(19)15(17)20-3-5-22(2,26)6-4-20;/h7-8,10H,3-6,9H2,1-2H3,(H,24,25);1H |
| SMIL | Cl.CC1COc2c(N3CC[N+](C)([O-])CC3)c(F)cc4C(=O)C(=CN1c24)C(=O)O |
| Molekylvægt (g/mol) | 413.83 |
| Vis mere |