missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
4-[(RS)-(4-Hydroxyphenyl)(pyridin-2-yl)methyl]phenyl Sodium Sulphate (4-[(Pyridin-2-yl)(4-hydroxyphenyl)methyl]phenyl Sodium Sulphate), Mikromol™
Beskrivelse
4-[(RS)-(4-Hydroxyphenyl)(pyridin-2-yl)methyl]phenyl Sodium Sulphate (4-[(Pyridin-2-yl)(4-hydroxyphenyl)methyl]phenyl Sodium Sulphate)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 4-[(RS)-(4-Hydroxyphenyl)(pyridin-2-yl)methyl]phenyl Sodium Sulfate (4-[(Pyridin-2-yl)(4-hydroxyphenyl)methyl]phenyl Sodium Sulfate) |
| CAS | 32500-19-9 |
| Analyt- eller komponentnavne | 4-[(RS)-(4-Hydroxyphenyl)(pyridin-2-yl)methyl]phenyl Sodium Sulfate (4-[(Pyridin-2-yl)(4-hydroxyphenyl)methyl]phenyl Sodium Sulfate) |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C18 H14 N O5 S . Na |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Sodium Picosulfate Imp. A (EP), Sodium Picosulfate USP Related Compound A, Sodium Picosulfate USP RC A, 4-[(RS)-(4-Hydroxyphenyl)(pyridin-2-yl)methyl]phenyl Sodium Sulphate, 4-[(Pyridin-2-yl)(4-hydroxyphenyl)methyl]phenyl Sodium Sulphate, Picosulfate Related Compound A as Sodium Salt, Sodium Picosulfate Impurity A, Sodium Picosulfate Impurity A as Sodium Salt |
| InChI formel | InChI=1S/C18H15NO5S.Na/c20-15-8-4-13(5-9-15)18(17-3-1-2-12-19-17)14-6-10-16(11-7-14)24-25(21,22)23;/h1-12,18,20H,(H,21,22,23);/q;+1/p-1 |
| SMIL | [Na+].Oc1ccc(cc1)C(c2ccc(OS(=O)(=O)[O-])cc2)c3ccccn3 |
| IUPAC navn | sodium;[4-[(4-hydroxyphenyl)-pyridin-2-ylmethyl]phenyl] sulfate |
| Vis mere |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?