Få mere at vide
4-[(RS)-(4-Hydroxyphenyl)-(pyridin-2-yl)methyl]phenyl Acetate, Mikromol™
Artikelnummer. 17941586
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17941586 100 mg
17931706 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17941586 Leverandør Mikromol Leverandørnr. MM0031.02
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-[(RS)-(4-Hydroxyphenyl)-(pyridin-2-yl)methyl]phenyl Acetate
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-[(RS)-(4-Hydroxyphenyl)-(pyridin-2-yl)methyl]phenyl Acetate
CAS 72901-16-7
Analyt- eller komponentnavne 4-[(RS)-(4-Hydroxyphenyl)-(pyridin-2-yl)methyl]phenyl Acetate
Forsendelsestilstand Room Temperature
Molekylær formel C20 H17 N O3
Certificeringer/compliance ISO 17025
Synonym Bisacodyl Imp. C (EP), Bisacodyl USP Related Compound C, Bisacodyl USP RC C, 4-[(RS)-(4-Hydroxyphenyl)-(pyridin-2-yl)methyl]phenyl Acetate, Bisacodyl Related Compound C, Bisacodyl Impurity C
InChI formel InChI=1S/C20H17NO3/c1-14(22)24-18-11-7-16(8-12-18)20(19-4-2-3-13-21-19)15-5-9-17(23)10-6-15/h2-13,20,23H,1H3
SMIL CC(=O)Oc1ccc(cc1)C(c2ccc(O)cc2)c3ccccn3
IUPAC navn [4-[(4-hydroxyphenyl)-pyridin-2-ylmethyl]phenyl] acetate
Molekylvægt (g/mol) 319.35
Mængde 100 mg
Formel vægt 319.1208 g/mol