Få mere at vide
4-(Pyrrolidin-1-yl)-1-(4-hydroxy-2,6-dimethoxyphenyl)butan-1-one Hydrochloride (p-Desmethyl-Buflomedil Hydrochloride), Mikromol™
Artikelnummer. 30049794
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Mængde
30049794 100 mg
30049409 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30049794 missing translation for 'mfr' Mikromol missing translation for 'supplierNo' MM0066.02
Denna artikel kan inte returneras. Se returpolicy
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-(Pyrrolidin-1-yl)-1-(4-hydroxy-2,6-dimethoxyphenyl)butan-1-one Hydrochloride (p-Desmethyl-Buflomedil Hydrochloride)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemisk navn eller materiale 4-(Pyrrolidin-1-yl)-1-(4-hydroxy-2,6-dimethoxyphenyl)butan-1-one Hydrochloride (p-Desmethyl-Buflomedil Hydrochloride)
CAS 70585-59-0
Analyt- eller komponentnavne 4-(Pyrrolidin-1-yl)-1-(4-hydroxy-2,6-dimethoxyphenyl)butan-1-one Hydrochloride (p-Desmethyl-Buflomedil Hydrochloride)
Forsendelsestilstand Room Temperature
Molekylær formel C16 H23 N O4 . Cl H
Certificeringer/compliance ISO 17025
Synonym Buflomedil Hydrochloride Imp. B (EP), Buflomedil Imp. B (EP), 4-(Pyrrolidin-1-yl)-1-(4-hydroxy-2,6-dimethoxyphenyl)butan-1-one Hydrochloride, Buflomedil Hydrochloride Imp. B (EP) as Hydrochloride, p-Desmethyl-Buflomedil Hydrochloride, Buflomedil Hydrochloride Impurity B as Hydrochloride, Buflomedil Impurity B as Hydrochloride
InChI formel InChI=1S/C16H23NO4.ClH/c1-20-14-10-12(18)11-15(21-2)16(14)13(19)6-5-9-17-7-3-4-8-17;/h10-11,18H,3-9H2,1-2H3;1H
SMIL Cl.COc1cc(O)cc(OC)c1C(=O)CCCN2CCCC2
IUPAC navn 1-(4-hydroxy-2,6-dimethoxyphenyl)-4-pyrrolidin-1-ylbutan-1-one;hydrochloride
Molekylvægt (g/mol) 329.82
Mængde 100 mg
Formel vægt 329.1394 g/mol
Visa mer Visa mindre
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.