Få mere at vide
4-(Pyrrolidin-1-yl)-1-(4-hydroxy-2,6-dimethoxyphenyl)butan-1-one Hydrochloride (p-Desmethyl-Buflomedil Hydrochloride), Mikromol™
Beskrivelse
4-(Pyrrolidin-1-yl)-1-(4-hydroxy-2,6-dimethoxyphenyl)butan-1-one Hydrochloride (p-Desmethyl-Buflomedil Hydrochloride)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 4-(Pyrrolidin-1-yl)-1-(4-hydroxy-2,6-dimethoxyphenyl)butan-1-one Hydrochloride (p-Desmethyl-Buflomedil Hydrochloride) |
| CAS | 70585-59-0 |
| Analyt- eller komponentnavne | 4-(Pyrrolidin-1-yl)-1-(4-hydroxy-2,6-dimethoxyphenyl)butan-1-one Hydrochloride (p-Desmethyl-Buflomedil Hydrochloride) |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C16 H23 N O4 . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Buflomedil Hydrochloride Imp. B (EP), Buflomedil Imp. B (EP), 4-(Pyrrolidin-1-yl)-1-(4-hydroxy-2,6-dimethoxyphenyl)butan-1-one Hydrochloride, Buflomedil Hydrochloride Imp. B (EP) as Hydrochloride, p-Desmethyl-Buflomedil Hydrochloride, Buflomedil Hydrochloride Impurity B as Hydrochloride, Buflomedil Impurity B as Hydrochloride |
| InChI formel | InChI=1S/C16H23NO4.ClH/c1-20-14-10-12(18)11-15(21-2)16(14)13(19)6-5-9-17-7-3-4-8-17;/h10-11,18H,3-9H2,1-2H3;1H |
| SMIL | Cl.COc1cc(O)cc(OC)c1C(=O)CCCN2CCCC2 |
| IUPAC navn | 1-(4-hydroxy-2,6-dimethoxyphenyl)-4-pyrrolidin-1-ylbutan-1-one;hydrochloride |
| Vis mere |