Få mere at vide
4-(Pyrrolidin-1-yl)-1-(2-hydroxy-4,6-dimethoxyphenyl)butan-1-one Hydrochloride (o-Desmethyl-Buflomedil Hydrochloride), Mikromol™
Beskrivelse
4-(Pyrrolidin-1-yl)-1-(2-hydroxy-4,6-dimethoxyphenyl)butan-1-one Hydrochloride (o-Desmethyl-Buflomedil Hydrochloride)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 4-(Pyrrolidin-1-yl)-1-(2-hydroxy-4,6-dimethoxyphenyl)butan-1-one Hydrochloride (o-Desmethyl-Buflomedil Hydrochloride) |
| CAS | 70585-57-8 |
| Analyt- eller komponentnavne | 4-(Pyrrolidin-1-yl)-1-(2-hydroxy-4,6-dimethoxyphenyl)butan-1-one Hydrochloride (o-Desmethyl-Buflomedil Hydrochloride) |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C16 H23 N O4 . Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Buflomedil Hydrochloride Imp. A (EP), Buflomedil Imp. A (EP), 4-(Pyrrolidin-1-yl)-1-(2-hydroxy-4,6-dimethoxyphenyl)butan-1-one Hydrochloride, Buflomedil Hydrochloride Imp. A (EP) as Hydrochloride, o-Desmethyl-Buflomedil Hydrochloride, Buflomedil Hydrochloride Impurity A as Hydrochloride, Buflomedil Impurity A as Hydrochloride |
| InChI formel | InChI=1S/C16H23NO4.ClH/c1-20-12-10-14(19)16(15(11-12)21-2)13(18)6-5-9-17-7-3-4-8-17;/h10-11,19H,3-9H2,1-2H3;1H |
| SMIL | Cl.COc1cc(O)c(C(=O)CCCN2CCCC2)c(OC)c1 |
| IUPAC navn | 1-(2-hydroxy-4,6-dimethoxyphenyl)-4-pyrrolidin-1-ylbutan-1-one;hydrochloride |
| Vis mere |