Få mere at vide
4-Pyridoxic Acid, TRC
Artikelnummer. 30009993
Skift visning
Mængde:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30009993 100 mg
30003642 1 g
30004416 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30009993 Leverandør Toronto Research Chemicals Leverandørnr. TRCP991875100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 82-82-6
Molekylær formel C8 H9 N O4
Molekylvægt (g/mol) 183.16
Synonym 4-Pyridoxic Acid, 3-Hydroxy-5-(hydroxymethyl)-2-methyl-4-pyridinecarboxylic Acid, 3-Hydroxy-5-(hydroxymethyl)-2-methyl-isonicotinic Acid, 2-Methyl-3-hydroxy-4-carboxy-5-hydroxymethylpyridine, 3-Hydroxy-5-(hydroxymethyl)-2-methylisonicotinic Acid, Pyridoxic Acid, 4-Pyridoxinecarboxylic Acid, 4-Pyridoxinic Acid, 4-Pyridoxylic Acid, NSC 41814
IUPAC navn 3-hydroxy-5-(hydroxymethyl)-2-methylpyridine-4-carboxylic acid
SMIL Cc1ncc(CO)c(C(=O)O)c1O
Tekniske data
Kemisk navn eller materiale 4-Pyridoxic Acid
Renhedsgrad noter HPLC
Mængde 100 mg
InChI formel InChI=1S/C8H9NO4/c1-4-7(11)6(8(12)13)5(3-10)2-9-4/h2,10-11H,3H2,1H3,(H,12,13)
Formel vægt 183.0532
Procent renhed >95%