Få mere at vide
4-Pyridoxic Acid-d3, TRC
Artikelnummer. 30422106
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30422106 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30422106 Leverandør Toronto Research Chemicals Leverandørnr. TRCP99187710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Pyridoxic Acid-d3
CAS 1435934-03-4
Renhedsgrad noter HPLC
Molekylær formel C8H6D3NO4
Mængde 10 mg
Synonym 3-Hydroxy-5-(hydroxymethyl)-2-methyl-4-pyridinecarboxylic Acid-d3, 3-Hydroxy-5-(hydroxymethyl)-2-methyl-isonicotinic Acid-d3, 2-Methyl-3-hydroxy-4-carboxy-5-hydroxymethylpyridine-d3, 3-Hydroxy-5-(hydroxymethyl)-2-methylisonicotinic Acid-d3, Pyridoxic Acid-d3, 4-Pyridoxinecarboxylic Acid-d3, 4-Pyridoxinic Acid-d3, 4-Pyridoxylic Acid-d3, NSC 41814-d3
InChI formel InChI=1S/C8H9NO4/c1-4-7(11)6(8(12)13)5(3-10)2-9-4/h2,10-11H,3H2,1H3,(H,12,13)/i1D3
SMIL OC1=C(C([2H])([2H])[2H])N=CC(CO)=C1C(O)=O
Molekylvægt (g/mol) 186.18
Formel vægt 186.072
Procent renhed >95