Få mere at vide
4-(Propylthio)-1,2-benzenediamine, TRC
Artikelnummer. 30412878
Skift visning
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30412878 500 mg
30423015 1 g
30410907 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30412878 Leverandør Toronto Research Chemicals Leverandørnr. TRCP838470500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 66608-52-4
Molekylær formel C9H14N2S
Molekylvægt (g/mol) 182.29
Synonym 4-(Propylthio)-1,2-phenylenediamine, 4-Propylthio-o-phenylenediamine
IUPAC navn 4-propylsulfanylbenzene-1,2-diamine
SMIL CCCSc1ccc(N)c(N)c1
Tekniske data
Kemisk navn eller materiale 4-(Propylthio)-1,2-benzenediamine
Mængde 500 mg
InChI formel InChI=1S/C9H14N2S/c1-2-5-12-7-3-4-8(10)9(11)6-7/h3-4,6H,2,5,10-11H2,1H3
Formel vægt 182.0878