Få mere at vide
4-Pregnen-21-ol-3,20-dione-2,2,4,6,6,17alpha,21,21-d8, CDN
Artikelnummer. 30318987
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30318987 0.005 g
30317731 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30318987 Leverandør CDN Leverandørnr. CDND57320.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Deoxy Corticosterone-d8
CAS 55487-63-3
Molekylær formel C21 H22 D8 O3
Mængde 0.005 g
InChI formel InChI=1 S/C21H30O3/c1-20-9-7-14(23)11-13(20)3-4-15-16-5-6-18(19(24)12-22)21(16,2)10-8-17(15)20/h11,15-18,22 H,3-10,12H2,1-2H3/t15-,16-,17-,18+,20-,21-/m0/s1/i3D2,7D2,11 D,12D2,18 D
SMIL [2 H]C1=C2C([2 H])([2 H])C[C@@H]3 [C@H](CC[C@@]4(C)[C@H]3 CC[C@]4([2 H])C(=O)C([2 H])([2 H])O)[C@@]2(C)CC([2 H])([2 H])C1=O
IUPAC navn (8 S,9 S,10 R,13 S,14 S,17 S)-2,2,4,6,6,17-hexadeuterio-17-(2,2-dideuterio-2-hydroxyacetyl)-10,13-dimethyl-7,8,9,11,12,14,15,16-octahydro-1 H-cyclopenta[a]phenanthren-3-one
Molekylvægt (g/mol) 338.51
Formel vægt 338.2697 g/mol
Procent renhed 96 atom % D, min 97% Chemical Purity