Få mere at vide
4-Pregnen-17alpha-ol-3,20-dione-2,2,4,6,6,21,21,21-d8, CDN
Artikelnummer. 30315342
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30315342 0.01 g
30315259 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30315342 Leverandør CDN Leverandørnr. CDND56500.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 17 A-Hydroxyprogesterone-D8 (2,2,4,6,6,21,21,21-D8)
CAS 850023-80-2
Molekylær formel C21 2H8 H22 O3
Mængde 0.01 g
Synonym Pregn-4-ene-3,20-dione-2,2,4,6,6,21,21,21-d8, 17-hydroxy-, 17α-Hydroxyprogesterone-D8 (2,2,4,6,6,21,21,21-D8), 2,2,4,6,6,21,21,21-d8-17α-Hydroxyprogesterone
InChI formel InChI=1 S/C21H30O3/c1-13(22)21(24)11-8-18-16-5-4-14-12-15(23)6-9-19(14,2)17(16)7-10-20(18,21)3/h12,16-18,24 H,4-11H2,1-3H3/t16-,17+,18+,19+,20+,21+/m1/s1/i1D3,4D2,6D2,12 D
SMIL [2 H]C1=C2C([2 H])([2 H])C[C@@H]3 [C@H](CC[C@@]4(C)[C@H]3 CC[C@]4(O)C(=O)C([2 H])([2 H])[2 H])[C@@]2(C)CC([2 H])([2 H])C1=O
IUPAC navn (8 R,9 S,10 R,13 S,14 S,17 R)-2,2,4,6,6-pentadeuterio-17-hydroxy-10,13-dimethyl-17-(2,2,2-trideuterioacetyl)-7,8,9,11,12,14,15,16-octahydro-1 H-cyclopenta[a]phenanthren-3-one
Molekylvægt (g/mol) 338.51
Formel vægt 338.2697 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity