Få mere at vide
4-Piperazin-1-yl-quinoline Dihydrochloride, TRC
Artikelnummer. 30419182
Skift visning
Mængde:
25 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30419182 250 mg
30420537 50 mg
30423124 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30419182 Leverandør Toronto Research Chemicals Leverandørnr. TRCP475448250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1350361-89-5
Molekylær formel C13H15N3.2[HCl]
Molekylvægt (g/mol) 286.2
Synonym Quinoline, 4-(1-piperazinyl)-, hydrochloride (1:2) (ACI), 4-Piperazin-1-yl-quinoline dihydrochloride, 4-(1-Piperazinyl)quinoline dihydrochloride, 4-(Piperazin-1-yl)quinoline dihydrochloride
IUPAC navn 4-piperazin-1-ylquinoline;dihydrochloride
SMIL Cl.Cl.C1CN(CCN1)c2ccnc3ccccc23
Tekniske data
Kemisk navn eller materiale 4-Piperazin-1-yl-quinoline Dihydrochloride
Mængde 250 mg
InChI formel InChI=1S/C13H15N3.2ClH/c1-2-4-12-11(3-1)13(5-6-15-12)16-9-7-14-8-10-16;;/h1-6,14H,7-10H2;2*1H
Formel vægt 285.08