Få mere at vide
4-Phthalimidoacetoacetic Acid Ethyl Ester, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 4-Phthalimidoacetoacetic Acid Ethyl Ester |
| CAS | 13855-80-6 |
| Molekylær formel | C14H13NO5 |
| Mængde | 10 mg |
| Synonym | Ethyl (phthalimidoacetyl)acetate, Ethyl 4-(1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)-3-oxobutanoate, Ethyl 4-phthalimidoacetoacetate, 4-Phthalimidoacetoacetic acid ethyl ester, 2-Isoindolineacetoacetic acid','1,3-dioxo-','ethyl ester (7CI','8CI), Ethyl 1,3-dihydro-β,1,3-trioxo-2H-isoindole-2-butanoate (ACI), Ethyl (phthalimidoacetyl)acetate, Ethyl 4-(1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)-3-oxobutanoate, Ethyl 4-phthalimidoacetoacetate |
| InChI formel | InChI=1S/C14H13NO5/c1-2-20-12(17)7-9(16)8-15-13(18)10-5-3-4-6-11(10)14(15)19/h3-6H,2,7-8H2,1H3 |
| SMIL | O=C(OCC)CC(=O)CN1C(=O)C=2C=CC=CC2C1=O |
| IUPAC navn | 2H-Isoindole-2-butanoic acid','1,3-dihydro-β,1,3-trioxo-','ethyl ester |
| Molekylvægt (g/mol) | 275.26 |
| Formel vægt | 275.08 |