Få mere at vide
4-Phospho D-Erythronate, TRC
Artikelnummer. 17932735
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17932735 10 mg
17902225 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17932735 Leverandør Toronto Research Chemicals Leverandørnr. TRCP35500010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 57229-25-1
Molekylær formel C4H9O8P
Molekylvægt (g/mol) 216.08
Synonym (2R,3R)-2,3-Dihydroxy-4-(phosphonooxy)butanoic Acid, [R-(R*,R*)]-2,3-Dihydroxy-4-(phosphonooxy)butanoic Acid;
SMIL C([C@H]([C@H](C(=O)O)O)O)OP(=O)(O)O
Tekniske data
Kemisk navn eller materiale 4-Phospho D-Erythronate
Mængde 10 mg
InChI formel InChI=1S/C4H9O8P/c5-2(3(6)4(7)8)1-12-13(9,10)11/h2-3,5-6H,1H2,(H,7,8)(H2,9,10,11)/t2-,3-/m1/s1
Formel vægt 216.0035