Få mere at vide
4-Phenylbutyric-d11 Acid, CDN
Artikelnummer. 30320403
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30320403 0.5 g 0.5g
30319879 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320403 Leverandør CDN Leverandørnr. CDND54990.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Phenylbutyric acid-d11
CAS 358730-86-6
Molekylær formel C10 2H11 H O2
Mængde 0.5 g
InChI formel InChI=1 S/C10H12O2/c11-10(12)8-4-7-9-5-2-1-3-6-9/h1-3,5-6 H,4,7-8H2,(H,11,12)/i1D,2 D,3 D,4D2,5 D,6 D,7D2,8D2
SMIL [2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)O
IUPAC navn 2,2,3,3,4,4-hexadeuterio-4-(2,3,4,5,6-pentadeuteriophenyl)butanoic acid
Molekylvægt (g/mol) 175.27
Formel vægt 175.1528 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity