Få mere at vide
4-(Phenyl-d5)-3-buten-2-one, TRC
Artikelnummer. 30424188
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30424188 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30424188 Leverandør Toronto Research Chemicals Leverandørnr. TRCP31958250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-(Phenyl-d5)-3-buten-2-one
CAS 56187-93-0
Molekylær formel C10 2H5 H5 O
Mængde 50 mg
InChI formel InChI=1S/C10H10O/c1-9(11)7-8-10-5-3-2-4-6-10/h2-8H,1H3/b8-7+/i2D,3D,4D,5D,6D
SMIL [2H]c1c([2H])c([2H])c(\C=C\C(=O)C)c([2H])c1[2H]
IUPAC navn (E)-4-(2,3,4,5,6-pentadeuteriophenyl)but-3-en-2-one
Molekylvægt (g/mol) 151.22
Formel vægt 151.1045