Learn More
4-Phenyl-5-(trifluoromethyl)thiophene-2-carboxylic acid, 95%, Thermo Scientific™
Product Code. 10644603
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10644603 1 g Amber Glass Bottle 1g
10149602 250 mg Amber Glass Bottle 250mg
10215334 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10644603 Supplier Thermo Scientific Maybridge Supplier No. SEW02660DA
 Restricted Item
Forventet forsendelse: 04-01-2027
Add to basket
Chemical Identifiers
CAS 208108-76-3
Molecular Formula C12H7F3O2S
Molecular Weight (g/mol) 272.241
InChI Key GIUAGKGTDDIMHB-UHFFFAOYSA-N
Synonym 4-phenyl-5-trifluoromethyl thiophene-2-carboxylic acid, 4-phenyl-5-trifluoromethyl-2-thiophenecarboxylic acid, 2-thiophenecarboxylic acid, 4-phenyl-5-trifluoromethyl, 2-thiophenecarboxylicacid, 4-phenyl-5-trifluoromethyl, maybridge3_006560, 4-phenyl-5-trifluoromethyl-thiophene-2-carboxylic acid, 4-phenyl-5-trifluoromethyl-2-thiophene carboxylic acid
PubChem CID 2783372
IUPAC Name 4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylic acid
SMILES C1=CC=C(C=C1)C2=C(SC(=C2)C(=O)O)C(F)(F)F
Specifications
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 272.25
Percent Purity 95%
Chemical Name or Material 4-Phenyl-5-(trifluoromethyl)thiophene-2-carboxylic acid