Få mere at vide
4-phenyl-2,3-dihydroquinazolin-2-one, TRC
Artikelnummer. 30410245
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30410245 50 mg
30415192 500 mg
30417596 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30410245 Leverandør Toronto Research Chemicals Leverandørnr. TRCP32294350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 23441-75-0
Molekylær formel C14H10N2O
Molekylvægt (g/mol) 222.25
Synonym 4-Phenylquinazolin-2(1H)-One
IUPAC navn 4-phenylquinazolin-2(1H)-one
SMIL O=C1N=C(C2=C(N1)C=CC=C2)C3=CC=CC=C3
Tekniske data
Kemisk navn eller materiale 4-phenyl-2,3-dihydroquinazolin-2-one
Mængde 50 mg
InChI formel InChI=1S/C14H10N2O/c17-14-15-12-9-5-4-8-11(12)13(16-14)10-6-2-1-3-7-10/h1-9H,(H,15,16,17)
Formel vægt 22.08