missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-1,2,3-thiadiazole-5-carboxylic acid, 97%, Thermo Scientific™

Catalog Number p-2702118
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
78875-63-5
C9H6N2O2S
206.219
MFCD00052216
DQZLGXDVESJWKA-UHFFFAOYSA-N
4-phenyl-1,2,3-thiadiazole-5-carboxylic acid, 1,2,3-thiadiazole-5-carboxylic acid, 4-phenyl, 4-phenyl-5-thiadiazolecarboxylic acid, 5-carboxy-4-phenyl-1,2,3-thiadiazole, 4-phenyl-1,2,3-thiadiazol-5-carboxylic acid, 4-phenyl 1,2,3 thiadiazole-5-carboxylic acid, 1,2,3-thiadiazole-5-carboxylicacid, 4-phenyl, 4-phenyl-1,2,3 thiadiazole-5-carboxylic acid
2737167
4-phenylthiadiazole-5-carboxylic acid
C1=CC=C(C=C1)C2=C(SN=N2)C(=O)O
This item is not returnable. View return policy
78875-63-5
C9H6N2O2S
206.219
MFCD00052216
DQZLGXDVESJWKA-UHFFFAOYSA-N
4-phenyl-1,2,3-thiadiazole-5-carboxylic acid, 1,2,3-thiadiazole-5-carboxylic acid, 4-phenyl, 4-phenyl-5-thiadiazolecarboxylic acid, 5-carboxy-4-phenyl-1,2,3-thiadiazole, 4-phenyl-1,2,3-thiadiazol-5-carboxylic acid, 4-phenyl 1,2,3 thiadiazole-5-carboxylic acid, 1,2,3-thiadiazole-5-carboxylicacid, 4-phenyl, 4-phenyl-1,2,3 thiadiazole-5-carboxylic acid
2737167
4-phenylthiadiazole-5-carboxylic acid
C1=CC=C(C=C1)C2=C(SN=N2)C(=O)O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 206.22
Percent Purity 97%
Chemical Name or Material 4-Phenyl-1, 2, 3-thiadiazole-5-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'