missing translation for 'onlineSavingsMsg'
Learn More

4-Phenoxybenzeneboronic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-7035236
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
51067-38-0
C12H11BO3
214.03
MFCD00093312
KFXUHRXGLWUOJT-UHFFFAOYSA-N
4-phenoxyphenyl boronic acid, 4-phenoxybenzeneboronic acid, 4-phenoxyphenyl boranediol, boronic acid, 4-phenoxyphenyl, p-phenoxyphenyl boronic acid, pubchem7890, acmc-209krn, phenoxyphenylboronic acid, d0d2ja
2734377
(4-phenoxyphenyl)boronic acid
OB(O)C1=CC=C(OC2=CC=CC=C2)C=C1
This item is not returnable. View return policy
51067-38-0
C12H11BO3
214.03
MFCD00093312
KFXUHRXGLWUOJT-UHFFFAOYSA-N
4-phenoxyphenyl boronic acid, 4-phenoxybenzeneboronic acid, 4-phenoxyphenyl boranediol, boronic acid, 4-phenoxyphenyl, p-phenoxyphenyl boronic acid, pubchem7890, acmc-209krn, phenoxyphenylboronic acid, d0d2ja
2734377
(4-phenoxyphenyl)boronic acid
OB(O)C1=CC=C(OC2=CC=CC=C2)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 141°C to 145°C
Quantity 1 g
Formula Weight 214.03
Percent Purity 98%
Chemical Name or Material 4-Phenoxybenzeneboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'