Få mere at vide
4-Penten-1-amine Hydrochloride, TRC
Artikelnummer. 30179022
Skift visning
Mængde:
1 g
2.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179022 2.5 g
30179587 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30179022 Leverandør Toronto Research Chemicals Leverandørnr. TRCP2274032.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 27546-60-7
Molekylær formel C5H11N • HCl
Molekylvægt (g/mol) 85.153646
Synonym 4-Pentenylamine Hydrochloride, 1-Amino-4-pentene Hydrochloride, 4-Penten-1-ylamine Hydrochloride, 4-Pentenamine Hydrochloride, 5-Aminopent-1-ene Hydrochloride
IUPAC navn pent-4-en-1-amine;hydrochloride
SMIL C=CCCCN.Cl
Tekniske data
Kemisk navn eller materiale 4-Penten-1-amine Hydrochloride
Mængde 2.5 g
InChI formel InChI=1S/C5H11N.ClH/c1-2-3-4-5-6;/h2H,1,3-6H2;1H
Formel vægt 121.0658