missing translation for 'onlineSavingsMsg'
Learn More

4-(Pentafluorothio)benzyl alcohol, 97%, Thermo Scientific™

Catalog Number p-7032784
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
773872-73-4
C7H7F5OS
234.184
MFCD06203063
QKEQGUCFBIRBAU-UHFFFAOYSA-N
4-pentafluorosulfur benzyl alcohol, 4-pentafluoro-??-sulfanyl phenyl methanol, 4-pentafluorothio benzyl alcohol
58360645
[4-(pentafluoro-$l^{6}-sulfanyl)phenyl]methanol
C1=CC(=CC=C1CO)S(F)(F)(F)(F)F
This item is not returnable. View return policy
773872-73-4
C7H7F5OS
234.184
MFCD06203063
QKEQGUCFBIRBAU-UHFFFAOYSA-N
4-pentafluorosulfur benzyl alcohol, 4-pentafluoro-??-sulfanyl phenyl methanol, 4-pentafluorothio benzyl alcohol
58360645
[4-(pentafluoro-$l^{6}-sulfanyl)phenyl]methanol
C1=CC(=CC=C1CO)S(F)(F)(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 234.18
Percent Purity 97%
Chemical Name or Material 4-(Pentafluorothio)benzyl alcohol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'